2',6'-Dihydroxyacetophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(2,6-Dihydroxyphenyl)ethan-1-one1-(2,6-Dihydroxyphenyl)ethanone

075425-100G 075425-25G
102664-046EA 24.62 USD
102664-046 102541-296
2',6'-Dihydroxyacetophenone ≥95%
2',6'-Dihydroxyacetophenone

Hazard: Irritant

Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 156…159 °C
MDL Number: MFCD00002270
CAS Number: 699-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR