3-Pyridinesulfonic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Pyridine-3-sulphonic acid

075422-10G 075422-100G
102541-284EA 64.62 USD
102541-284 102541-286
3-Pyridinesulfonic acid ≥95%
3-Pyridinesulphonic acid

Hazard: Irritant

Formula: C₅H₅NO₃S
MW: 159.16 g/mol
MDL Number: MFCD00006381
CAS Number: 636-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR