5-Bromo-3-nitro-o-toluic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2-methyl-3-nitrobenzoic acid
075418-25G
075418-5G
102833-826EA
509.23
USD
102833-826
102541-266
5-Bromo-3-nitro-o-toluic acid ≥95%
5-Bromo-3-nitro-o-toluic acid
Hazard: Irritant
Formula:
C₈H₆BrNO₄ MW: 260.0400 g/mol |
MDL Number:
MFCD07357316 CAS Number: 107650-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...