Methyl-4-methyl-3-nitrobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-methyl-3-nitrobenzoate

075414-100G
102541-246EA 73.85 USD
102541-246
Methyl-4-methyl-3-nitrobenzoate ≥95%
Methyl 4-methyl-3-nitrobenzoate

Hazard: Irritant

Formula: C₉H₉NO₄
MW: 195.18 g/mol
Melting Pt: 84…85 °C
MDL Number: MFCD00130054
CAS Number: 7356-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR