5-Hydroxysophthalic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Hydroxyisophthalic acid
075396-100G
075396-10G
102541-162EA
64.62
USD
102541-162
102541-164
5-Hydroxysophthalic acid ≥95%
5-Hydroxysophthalic acid
Hazard: Irritant
Formula:
C₈H₆O₅ MW: 182.13 g/mol |
MDL Number:
MFCD00002515 CAS Number: 618-83-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...