9-Acridone ≥95%
Supplier: Matrix Scientific
Synonyms:
Acridin-9(10H)-one
, 9(10H)-Acridone
075393-5G
102541-148EA
35.38
USD
102541-148
9-Acridone ≥95%
9-Acridone
Hazard: Irritant
Formula:
C₁₃H₉NO MW: 195.22 g/mol |
MDL Number:
MFCD00005019 CAS Number: 578-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...