N-Phenyl-p-toluidine ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Methyldiphenylamine
075390-100G
075390-10G
102541-130EA
143.08
USD
102541-130
102541-132
N-Phenyl-p-toluidine ≥95%
N-Phenyl-p-toluidine
Hazard: Irritant
Formula:
C₁₃H₁₃N MW: 183.25 g/mol |
MDL Number:
MFCD00092921 CAS Number: 620-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...