2,3-Dimethoxybenzoic acid ≥95%

Supplier: Matrix Scientific

075379-100G
102541-084EA 56.92 USD
102541-084
2,3-Dimethoxybenzoic acid ≥95%
2,3-Dimethoxybenzoic acid

Hazard: Irritant

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
MDL Number: MFCD00002432
CAS Number: 1521-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR