4-Amino-2,2,6,6-tetramethylpiperidine ≥95%
Supplier: Matrix Scientific
075353-100G
075353-10G
102540-968EA
278.46
USD
102540-968
102540-966
4-Amino-2,2,6,6-tetramethylpiperidine ≥95%
4-Amino-2,2,6,6-tetramethylpiperidine
Hazard: Irritant
Formula:
C₉H₂₀N₂ |
MDL Number:
MFCD00005984 CAS Number: 36768-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...