N-Ethylcyclohexylamine ≥95%

Supplier: Matrix Scientific

075350-100G 075350-10G
102540-952EA 307.69 USD
102540-952 102540-950
N-Ethylcyclohexylamine ≥95%
N-Ethylcyclohexylamine

Hazard: Irritant

Formula: C₈H₁₇N
MDL Number: MFCD00003834
CAS Number: 5459-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR