3-Chlorophthalic anhydride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloroisobenzofuran-1,3-dione

075346-100G 075346-25G
102540-938EA 129.23 USD
102540-938 102833-816
3-Chlorophthalic anhydride ≥95%
3-Chlorophthalic anhydride

Hazard: Irritant

Formula: C₈H₃ClO₃
MW: 182.56 g/mol
MDL Number: MFCD00023107
CAS Number: 117-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR