(3-Chlorophenyl)acetonitrile ≥95%
Supplier: Matrix Scientific
075338-10G
075338-100G
102540-902EA
293.85
USD
102540-902
102540-900
(3-Chlorophenyl)acetonitrile ≥95%
(3-Chlorophenyl)acetonitrile
Hazard: Toxic
Formula:
C₈H₆ClN MW: 151.6 g/mol Boiling Pt: 134…136 °C (10 mmHg) Melting Pt: 11…13 °C Density: 1.189 Flash Pt: >110 °C |
MDL Number:
MFCD00001908 CAS Number: 1529-41-5 UN: 3276 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...