1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) ≥95%

Supplier: Matrix Scientific

Synonyms: 1,4-Cyclohexanedicarboxylic acid

075305-100G
102540-762EA 100 USD
102540-762
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) ≥95%
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers)

Hazard: Irritant

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
MDL Number: MFCD00001465
CAS Number: 1076-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR