4-Fluorophenethylamine ≥95%

Supplier: Matrix Scientific

075296-100G
102540-720EA 149.23 USD
102540-720
4-Fluorophenethylamine ≥95%
4-Fluorophenethylamine

Hazard: Toxic/Corrosive

Formula: C₈H₁₀FN
Boiling Pt: 50…52 °C
Density: 1.061
Flash Pt: 79 °C
MDL Number: MFCD00134208
CAS Number: 1583-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR