2-Bromo-3,4-difluoroaniline ≥95%

Supplier: Matrix Scientific

075281-5G
102540-648EA 176.92 USD
102540-648
2-Bromo-3,4-difluoroaniline ≥95%
2-Bromo-3,4-difluoroaniline

Hazard: Toxic

Formula: C₆H₄BrF₂N
MW: 208.00 g/mol
MDL Number: MFCD11035924
CAS Number: 1092349-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR