2,1,3-Benzothiadiazole ≥95%

Supplier: Matrix Scientific

075273-100G
102540-608EA 83.95 USD
102540-608
2,1,3-Benzothiadiazole ≥95%
2,1,3-Benzothiadiazole

Hazard: Irritant

Formula: C₆H₄N₂S
Melting Pt: 44 °C
Flash Pt: 95 °C
MDL Number: MFCD00005809
CAS Number: 273-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR