8-Quinolinesulfonic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 8-Quinolinesulphonic acid8-Quinolinesulfonic acid

075272-10G 075272-100G
102540-606EA 346.15 USD
102540-606 102540-604
8-Quinolinesulfonic acid ≥95%
8-Quinolinesulphonic acid

Hazard: Irritant

Formula: C₉H₇NO₃S
MW: 209.23 g/mol
MDL Number: MFCD00024037
CAS Number: 85-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR