Methyl-4-fluoro-3-methylbenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-fluoro-3-methylbenzoate
075269-1G
075269-10G
075269-5G
102540-592EA
440
USD
102540-592
102540-590
102540-588
Methyl-4-fluoro-3-methylbenzoate ≥95%
Methyl 4-fluoro-3-methylbenzoate
Hazard: Irritant
Formula:
C₉H₉FO₂ MW: 168.1650 g/mol |
MDL Number:
MFCD06203783 CAS Number: 180636-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...