4'-Hydroxyheptanophenone ≥95%

Supplier: Matrix Scientific

075255-10G 075255-100G 075255-500G
102540-518EA 58.46 USD
102540-518 102540-522 102540-520
4'-Hydroxyheptanophenone ≥95%
4'-Hydroxyheptanophenone

Hazard: Irritant

Formula: C₁₃H₁₈O₂
MW: 206.2800 g/mol
MDL Number: MFCD00191523
CAS Number: 14392-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR