3,3-Dimethylbutyric acid ≥95%

Supplier: Matrix Scientific

075241-100G
102540-458EA 78.46 USD
102540-458
3,3-Dimethylbutyric acid ≥95%
3,3-Dimethylbutyric acid

Hazard: Corrosive

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 185…190 °C
Melting Pt: 6 °C
Density: 0.912
Flash Pt: 88 °C
MDL Number: MFCD00002715
CAS Number: 1070-83-3
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR