Methyl isobutyrylacetate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-methyl-3-oxopentanoate
075192-250G
075192-100G
102540-234EA
93.85
USD
102540-234
102664-038
Methyl isobutyrylacetate ≥95%
Methyl isobutyrylacetate
Hazard: Irritant
Formula:
C₇H₁₂O₃ MW: 144.17 g/mol Boiling Pt: 192 °C Density: 1.01 Flash Pt: 79 °C |
MDL Number:
MFCD00040499 CAS Number: 42558-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...