Hydroquinone monomethyl ether ≥95%

Supplier: Matrix Scientific

Synonyms: p-MethoxyphenolMequinol4-Methoxyphenol1-Hydroxy-4-methoxybenzene4-Hydroxyanisole

075183-100G
102540-188EA 60 USD
102540-188
Hydroquinone monomethyl ether ≥95%
Hydroquinone monomethyl ether

Hazard: Irritant

Formula: C₇H₈O₂
MW: 124.14 g/mol
MDL Number: MFCD00002332
CAS Number: 150-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR