4-Piperidone monohydrate hydrochloride ≥95%

Synonyms: Piperidin-4-one hydrate hydrochloride4-Azacyclohexanone Monohydrate Hydrochloride4,4-Dihydroxypiperidine Hydrochloride4-Piperidinone monohydrate hydrochloride4,4-Piperidinediol Hydrochloride4-Ketopiperidine Monohydrate Hydrochloride

075177-25G 075177-100G
102551-942EA 41.54 USD
102551-942 102540-158
4-Piperidone monohydrate hydrochloride ≥95%
4-Piperidone monohydrate hydrochloride

Hazard: Irritant

Formula: C₅H₉NO·xHCl·yH₂O
MW: 99.13 g/mol (free base)
Melting Pt: 94…100 °C
MDL Number: MFCD00012776
CAS Number: 40064-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR