2-Imidazolidinone ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Imidazolidone
075126-10G
075126-100G
102539-890EA
303.22
USD
102539-890
102539-892
2-Imidazolidinone ≥95%
2-Imidazolidinone
Hazard: Irritant
Formula:
C₃H₆N₂O MW: 86.0925 g/mol |
MDL Number:
MFCD00005257 CAS Number: 120-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...