2-Adamantanone ≥95%

Supplier: Matrix Scientific

Synonyms: Adamantan-2-one

075119-100G
102539-856EA 72.31 USD
102539-856
2-Adamantanone ≥95%
2-Adamantanone

Hazard: Irritant

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Melting Pt: 257 °C (subl.)
MDL Number: MFCD00074737
CAS Number: 700-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR