3-Isopropoxyphenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Isopropoxybenzeneboronic acid

075107-1G 075107-5G 075107-10G
102539-796EA 107.69 USD
102539-796 102539-798 102539-800
3-Isopropoxyphenylboronic acid ≥95%
3-Isopropoxyphenylboronic acid

Hazard: Irritant

Formula: C₉H₁₃BO₃
MW: 180.01 g/mol
MDL Number: MFCD03701512
CAS Number: 216485-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR