3-Amino-o-toluic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Amino-2-methylbenzoic acid
075106-100G
102539-794EA
120
USD
102539-794
3-Amino-o-toluic acid ≥95%
3-Amino-o-toluic acid
Hazard: Irritant
Formula:
C₈H₉NO₂ MW: 151.16 g/mol |
MDL Number:
MFCD00075026 CAS Number: 52130-17-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...