1-Phenylpiperazine ≥95%

Supplier: Matrix Scientific

Synonyms: N-Phenylpiperazine

075097-100G
102539-750EA 158.46 USD
102539-750
1-Phenylpiperazine ≥95%
1-Phenylpiperazine

Hazard: Irritant

Formula: C₁₀H₁₄N₂
MW: 162.23 g/mol
MDL Number: MFCD00005957
CAS Number: 92-54-6
UN: 2927
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR