N-Boc-L-phenylalaninol ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-Phenylalaninol(S)-(-)-BOC-Phenylalaninol

075086-5G
102539-694EA 42.55 USD
102539-694
N-Boc-L-phenylalaninol ≥95%
N-Boc-L-phenylalaninol

Hazard: Irritant

Formula: C₁₄H₂₁NO₃
MW: 251.3 g/mol
MDL Number: MFCD00076976
CAS Number: 66605-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR