2-Amino-5-nitrobenzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Nitroanthranilonitrile2-Cyano-4-nitroaniline

075063-100G
102539-574EA 138.46 USD
102539-574
2-Amino-5-nitrobenzonitrile ≥95%
2-Amino-5-nitrobenzonitrile

Hazard: Irritant

Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
Melting Pt: 207…210 °C
MDL Number: MFCD00007362
CAS Number: 17420-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR