N-Boc-D-alaninol ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-2-(BOC-amino)-1-propanolBoc-D-Alaninol

075048-100G
102539-492EA 267.69 USD
102539-492
N-Boc-D-alaninol ≥95%
N-Boc-D-alaninol

Hazard: Irritant

Formula: C₈H₁₇NO₃
MW: 175.2 g/mol
Melting Pt: 58…62 °C
MDL Number: MFCD00235912
CAS Number: 106391-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR