3-Bromo-2-chlorophenol ≥95%
Supplier: Matrix Scientific
075043-5G
075043-1G
102539-464EA
109.23
USD
102539-464
102539-466
3-Bromo-2-chlorophenol ≥95%
3-Bromo-2-chlorophenol
Hazard: Irritant
Formula:
C₆H₄BrClO MW: 207.452 g/mol |
MDL Number:
MFCD08166321 CAS Number: 863870-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...