1,3-Cyclohexanedione ≥95%

Supplier: Matrix Scientific

Synonyms: Cyclohexane-1,3-dione

075030-100G
102539-402EA 27.6 USD
102539-402
1,3-Cyclohexanedione ≥95%
1,3-Cyclohexanedione

Hazard: Irritant

Formula: C₆H₈O₂
MW: 112.13 g/mol
Melting Pt: 101…105 °C
MDL Number: MFCD00001585
CAS Number: 504-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR