2-Bromo-6-chloro-4-nitroaniline ≥95%

Supplier: Matrix Scientific

075019-10G 075019-100G
102539-350EA 36.92 USD
102539-350 102539-352
2-Bromo-6-chloro-4-nitroaniline ≥95%
2-Bromo-6-chloro-4-nitroaniline

Hazard: Irritant

Formula: C₆H₄BrClN₂O₂
MW: 251.47 g/mol
MDL Number: MFCD00014770
CAS Number: 99-29-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR