Ethyl (R)-(-)-3-hydroxybutyrate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl (R)-3-hydroxybutyrate
, (R)-(-)-3-Hydroxybutyric Acid Ethyl Ester
075002-25G
075002-100G
075002-5G
102664-024EA
156.92
USD
102664-024
102664-022
102539-270
Ethyl (R)-(-)-3-hydroxybutyrate ≥95%
Ethyl (R)-(-)-3-hydroxybutyrate
Purity: 95+%
CAS: 24915-95-5
MDL Number: MFCD00075386
Molecular Formula: C6H12O3
Molecular Weight: 132,16
Boiling Point: 75-76°C/12mmHg
Hazard: Irritant/Refrigerate
Formula:
C₆H₁₂O₃ MW: 132.16 g/mol Boiling Pt: 75…76 °C (12 mmHg) Density: 1.017 Flash Pt: 64 °C |
MDL Number:
MFCD00075386 CAS Number: 24915-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...