Cyclopentylamine ≥95%

Supplier: Matrix Scientific

Synonyms: Aminocyclopentane

074997-100G
102539-244EA 93.85 USD
102539-244
Cyclopentylamine ≥95%
Cyclopentylamine

Hazard: Flammable/Toxic/Corrosive

Formula: C₅H₁₁N
MW: 85.15 g/mol
Boiling Pt: 106…108 °C
Density: 0.863
Flash Pt: 13 °C
MDL Number: MFCD00001380
CAS Number: 1003-03-8
UN: 3286
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR