Nipecotamide ≥95%

Supplier: Matrix Scientific

Synonyms: Piperidine-3-carboxamide

074995-5G 074995-10G 074995-100G
102539-232EA 107.69 USD
102539-232 102539-234 102539-236
Nipecotamide ≥95%
Nipecotamide

Hazard: Irritant

Formula: C₆H₁₂N₂O
MW: 128.1700 g/mol
MDL Number: MFCD00005993
CAS Number: 4138-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR