3,4-Dihydro-2H-pyran ≥95%

Supplier: Matrix Scientific

074989-100G
102539-202EA 80 USD
102539-202
3,4-Dihydro-2H-pyran ≥95%
3,4-Dihydro-2H-pyran

Hazard: Flammable

Formula: C₅H₈O
Boiling Pt: 86 °C
Density: 0.922
Flash Pt: –16 °C
MDL Number: MFCD00006558
CAS Number: 110-87-2
UN: 2376
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR