Citrazinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Hydroxy-2-oxo-1,2-dihydropyridine-4-carboxylic acid2,6-Dihydroxyisonicotinic acid

074972-100G 074972-10G
102539-116EA 36.92 USD
102539-116 102539-118
Citrazinic acid ≥95%
Citrazinic acid

Hazard: Irritant

Formula: C₆H₅NO₄
MW: 155.11 g/mol
MDL Number: MFCD00006274
CAS Number: 99-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR