4-tert-Butylbenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(tert-Butyl)benzoic acid

074971-100G
102539-114EA 26.15 USD
102539-114
4-tert-Butylbenzoic acid ≥95%
4-tert-Butylbenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Melting Pt: 167 °C
MDL Number: MFCD00002563
CAS Number: 98-73-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR