5-Bromosalicylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-hydroxybenzoic acid

074964-100G
102539-080EA 223.08 USD
102539-080
5-Bromosalicylic acid ≥95%
5-Bromosalicylic acid

Hazard: Irritant

Formula: C₇H₅BrO₃
MW: 217.02 g/mol
MDL Number: MFCD00002455
CAS Number: 89-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR