2,5-Di-tert-butylhydroquinone ≥95%

Supplier: Matrix Scientific

Synonyms: BHQ

074963-10G 074963-100G
102539-074EA 36.92 USD
102539-074 102539-076
2,5-Di-tert-butylhydroquinone ≥95%
2,5-Di-tert-butylhydroquinone

Hazard: Irritant

Formula: C₁₄H₂₂O₂
MW: 222.33 g/mol
MDL Number: MFCD00008825
CAS Number: 88-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR