3,4-Dimethoxyaniline ≥95%

Supplier: Matrix Scientific

074941-100G
102538-966EA 115.38 USD
102538-966
3,4-Dimethoxyaniline ≥95%
3,4-Dimethoxyaniline

Purity: 95+%
CAS: 6315-89-5
MDL Number: MFCD00008394
Molecular Formula: C8H11NO2
Molecular Weight: 153,18
Hazard: Irritant-Harmful/Refrigerate/Light Sensitive

Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Melting Pt: 85…89 °C
MDL Number: MFCD00008394
CAS Number: 6315-89-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR