2-Methoxy-6-methylpyridine ≥95%
Supplier: Matrix Scientific
074940-10G
074940-100G
074940-5G
102538-962EA
493.35
USD
102538-962
102538-960
102538-958
2-Methoxy-6-methylpyridine ≥95%
2-Methoxy-6-methylpyridine
Hazard: Irritant
Formula:
C₇H₉NO MW: 123.16 g/mol |
MDL Number:
MFCD04112507 CAS Number: 63071-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...