N-(2-Bromoethyl)phthalimide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(2-Bromoethyl)isoindoline-1,3-dione

074933-10G 074933-100G
102538-928EA 23.08 USD
102538-928 102538-930
N-(2-Bromoethyl)phthalimide ≥95%
N-(2-Bromoethyl)phthalimide

Hazard: Irritant

Formula: C₁₀H₈BrNO₂
MW: 254.08 g/mol
MDL Number: MFCD00005902
CAS Number: 574-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR