(±)-Ethyl-2-bromopropionate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-bromopropionateDL-Ethyl 2-bromopropionate

074926-100G
102538-902EA 73.85 USD
102538-902
(±)-Ethyl-2-bromopropionate ≥95%
(±)-Ethyl 2-bromopropionate

Hazard: Irritant

Formula: C₅H₉BrO₂
MW: 181.03 g/mol
MDL Number: MFCD00000144
CAS Number: 535-11-5
UN: 1992
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR