4-Guanidinobenzoic acid hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: N-(4-Carboxyphenyl)guanidine hydrochloride

074905-5G
102538-786EA 49.23 USD
102538-786
4-Guanidinobenzoic acid hydrochloride ≥95%
4-Guanidinobenzoic acid hydrochloride

Hazard: Irritant

Formula: C₈H₁₀ClN₃O₂
MW: 215.64 g/mol
MDL Number: MFCD00040587
CAS Number: 42823-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR