6-Methoxyisatin ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Methoxyindoline-2,3-dione

074899-1G 074899-5G
102538-756EA 1535.38 USD
102538-756 102538-754
6-Methoxyisatin ≥95%
6-Methoxyisatin

Hazard: Irritant

Formula: C₉H₇NO₃
MW: 177.1568 g/mol
MDL Number: MFCD00667731
CAS Number: 52351-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR