4,6-Dichloropyridin-3-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4-Dichloro-5-hydroxypyridine

074895-1G
102538-738EA 530.77 USD
102538-738
4,6-Dichloropyridin-3-ol ≥95%
4,6-Dichloropyridin-3-ol

Hazard: Irritant

Formula: C₅H₃Cl₂NO
MW: 163.99 g/mol
MDL Number: MFCD13190072
CAS Number: 1196157-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR