(S)-(-)-4-Methoxy-α-methylbenzylamine ≥95%
Supplier: Matrix Scientific
Synonyms:
(S)-(-)-1-(4-Methoxyphenyl)ethylamine
, (S)-1-(4-Methoxyphenyl)ethylamine
074891-25G
074891-100G
102833-736EA
153.85
USD
102833-736
102538-722
(S)-(-)-4-Methoxy-α-methylbenzylamine ≥95%
(S)-(-)-4-Methoxy-α-methylbenzylamine
Hazard: Corrosive/Air Sensitive
Formula:
C₉H₁₃NO MW: 151.21 g/mol Boiling Pt: 65 °C (0.38 mmHg) Density: 1.02 |
MDL Number:
MFCD00671660 CAS Number: 41851-59-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...